Stainless Steel Featherweight Spring Forceps
Stainless Steel Featherweight Spring Forceps
Actual product may vary
Stainless Steel Featherweight Spring Forceps
Use these forceps for general laboratory applications. S Murray™ Stainless Steel Featherweight Spring Forceps are suitable for theatre, ward, veterinary, dental and chiropody use.
Have Questions?
Catalog NumberLength (Metric)
FKS12301379102 mm
Catalog number FKS12301379
Price (MYR)
234.00
PK
Add to cart
Length (Metric):
102 mm
Price (MYR)
234.00
PK
Add to cart
Use these forceps for general laboratory applications. S Murray™ Stainless Steel Featherweight Spring Forceps are suitable for theatre, ward, veterinary, dental and chiropody use.
  • Manufactured from high grade stainless steel
  • Complies with ISO 7153-1 specifications
  • Standard instruments are finished to a bright mirror polish
Specifications
Product TypeFeatherweight Spring Forceps
MaterialStainless Steel
For Use With (Application)For dissection
Quantity5/Pk.
Length (Metric)102 mm
Unit SizePK