Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Chemical Label | Description | Closure Color |
---|---|---|---|---|
FKS12196220 | 250 mL | Isopropanol | Printed in English, French, German and Spanish | Blue |
FKS12357512 | 500 mL | Ethanol | Region 2: Norwegian, Danish, Swedish, English | Orange |
FKS12550456 | 500 mL | Methanol | Printed in English, French, German and Spanish | Green |
FKS11303524 | 500 mL | Isopropanol | Printed in English, French, German and Spanish | Blue |