Pyrex™ Low-form Griffin Beakers
Pyrex™ Low-form Griffin Beakers
Actual product may vary
Fisherbrand
Pyrex™ Low-form Griffin Beakers
Measure liquids on bench top or hot plate without fear of chemical attack. Pyrex™ low-form Griffin beakers come in a wide variety of sizes with white gradations and marking spot.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1199651850 mL
FKS125342575 mL
FKS1196377910 mL
FKS11987564100 mL
FKS11997564250 mL
FKS119175741000 mL
FKS119265282000 mL
Catalog number FKS11996518
Price (MYR)
707.00
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
707.00
Add to cart
Measure liquids on bench top or hot plate without fear of chemical attack. Pyrex™ low-form Griffin beakers come in a wide variety of sizes with white gradations and marking spot.
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex™ borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • White graduations and marking spot
Specifications
Diameter (Metric) Outer42 mm
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Height (Metric)60 mm
Quantity10/Pk.
TypeBeaker
GraduatedYes
Certifications/ComplianceISO 3819, DIN 12331
ScaleWhite Graduated Scale
Unit Size