Pipette Tips: Low Retention
Pipette Tips: Low Retention
Actual product may vary
Pipette Tips: Low Retention
For use with Ovation and MLA Brand pipetters
Have Questions?
Catalog NumberFilteredSterilityVolume (Metric)
FKS11955992Non-filteredNon-sterile1250 μL
Catalog number FKS11955992
Price (MYR)
2,245.00
PK
Add to cart
Filtered:
Non-filtered
Sterility:
Non-sterile
Volume (Metric):
1250 μL
Price (MYR)
2,245.00
PK
Add to cart
  • Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
  • Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
DescriptionLow Retention
Volume (Metric)1250 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
Rack ColorBlue
AutoclavableAutoclavable
Length (English)2.2 in.
Thickness5 in.
For Use With (Application)Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications
Sterilization Methode-beam
CompatibilityMLA/Ovation Pipettes
FormatRack
Tip StyleLow Retention, Straight
PackagingVistaRak: 5 racks
Quantity192 Tips/Rack
Product TypePipette Tips
Color CodeBlue
DisposableSingle-use
For Use With (Equipment)MLA, Ovation
GraduatedNo
MaterialPolypropylene
ShapeCone
Unit SizePK