MBL™ Glass Graduated Cylinder with Polyethylene Stopper
MBL™ Glass Graduated Cylinder with Polyethylene Stopper
Actual product may vary
MBL™ Glass Graduated Cylinder with Polyethylene Stopper
Manufactured from borosilicate glass
Have Questions?
Catalog NumberCapacity (Metric)GraduationsTolerance
FKS11951518100 mL1 mL±1 mL
Catalog number FKS11951518
Price (MYR)
759.00
PK
Add to cart
Capacity (Metric):
100 mL
Graduations:
1 mL
Tolerance:
±1 mL
Price (MYR)
759.00
PK
Add to cart
Manufactured from borosilicate glass
  • Complies with ISO 4788
  • Supplied with polyethylene stopper
  • Graduations in easy to read blue enamel
  • Robust hexagonal foot
Specifications
Base TypeHexagonal
Scale ColorBlue
ShapeRound
TypeGraduated Cylinder
Tolerance±1 mL
MaterialBorosilicate Glass
Certifications/ComplianceDIN 12685, ISO 4788
ClassB
Capacity (Metric)100 mL
Height (Metric)291 mm
IncludesPolypropylene Stopper
ScaleBlue enamel
Stopper Number24/29
Stopper MaterialPolypropylene
Stopper StyleStopper
Closure MaterialPolyethylene
Graduations1 mL
Unit SizePK