Disposable Cuvettes
Disposable Cuvettes
Actual product may vary
Fisherbrand
Disposable Cuvettes
Choice of polystyrene or methacrylate
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)TypeMaterialWavelength Range
FKS119443854.5 mLStandard CuvettePMMA285 to 750 nm
FIS14-955-1294.5 mLCuvettePolystyrene340 to 750 nm
FKS119543954.5 mLCuvettePolystyrene340 to 750 nm
FIS14-955-1264.5 mLStandard CuvettePMMA285 to 750 nm
FIS14-955-1254.5 mLStandard CuvettePolystyrene340 to 750 nm
FKS116825994.5 mLStandard CuvettePolystyrene340 to 750 nm
FIS14-955-1271.5 mLSemi-micro CellPolystyrene340 to 750 nm
FKS116026091.5 mLSemi-micro CellPolystyrene340 to 750 nm
FIS14-955-1281.5 mLSemi-micro CellMethacrylate285 to 750 nm
Catalog number FKS11944385
Price (MYR)
537.00
Add to cart
Capacity (Metric):
4.5 mL
Type:
Standard Cuvette
Material:
PMMA
Wavelength Range:
285 to 750 nm
Price (MYR)
537.00
Add to cart
Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.
  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Choice of 4.5 and 1.5mL sample capacity
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
  • Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets
Specifications
TypeStandard Cuvette
DisposableSingle-use
Capacity (Metric)4.5 mL
MaterialPMMA
Length (Metric)12mm
Quantity500 Case
Path Length10 mm
Wavelength Range285 to 750 nm
Height (Metric)45 mm
For Use With (Equipment)Spectrophotometers, Fluorometers and Nephelometers
Width (Metric)12 mm
For Use With (Application)For assays in visible spectral range, accuracy throughout UV/Visible range
Unit Size