Pyrex™ Glass Graduated Cylinder, Class A
Pyrex™ Glass Graduated Cylinder, Class A
Actual product may vary
Fisherbrand
Pyrex™ Glass Graduated Cylinder, Class A
Manufactured from chemically resistant borosilicate glass. Pyrex™ Glass Graduated Cylinder, Class A is scaled with white enamel graduations for high visibility against liquids of any color. Constructed with a hexagonal base, the sturdy cylinders are available in a wide range of capacities.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS11933789100 mL1 mL
FKS11943789500 mL5 mL
Catalog number FKS11933789
Price (MYR)
484.00
Add to cart
Capacity (Metric):
100 mL
Graduations:
1 mL
Price (MYR)
484.00
Add to cart
Manufactured from chemically resistant borosilicate glass. Pyrex™ Glass Cylinders, Class A are scaled with white enamel graduations for high visibility against liquids of any color. Constructed with a hexagonal base, the sturdy cylinders are available in a wide range of capacities.
  • Features: sturdy hexagonal base, white ceramic markings, wide range of capacities
  • Class A: conformity certified to German calibration regulations, imprinted with conformity symbol “H” 
  • Batch certification: inscribed with individual serial number for traceability to national standards
  • Complies: ISO 4788
Specifications
Base TypeHexagonal
Scale ColorWhite
ShapeRound
TypeGraduated Cylinder
Tolerance±0.5 mL
MaterialBorosilicate Glass
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
ClassA
Capacity (Metric)100 mL
Height (Metric)260 mm
ScaleWhite enamel
Graduations1 mL
Unit Size