Designed for moisture determination balances. Fisherbrand™ Aluminum Weighing and Drying Dish is ideal for general purpose weighing, storage, processing or drying. Extra large, light-weight aluminum dishes measure 108mm diameter. Includes 50 disposable, stackable dishes per pack.
Have Questions?
Change view
Catalog Number
Size
Diameter (Metric)
Height (Metric)
FKS11908094
Medium
57 mm
12 mm
FKS11918094
Large
70 mm
16 mm
FKS11988084
Large
101 mm
8 mm
3 Options
Catalog number FKS11908094
Price (MYR)
90.00
Add to cart
Size:
Medium
Diameter (Metric):
57 mm
Height (Metric):
12 mm
Price (MYR)
90.00
Add to cart
Ideal for sample weighing, dispensing, storage, food processing, evaporating or drying
Also ideal as a dust cover for open vessels or as sample holders
Made from oil-free, light gauge aluminum that is FDA/food-grade compliant
Tapered walls promote stable stacking
Available in 4 varieties
Boats: 3 sizes with crimped sides, flat bottoms and finger tabs that can be etched for easy identification (100/pack)
Dish: 1 large size, designed for moisture determination balance (50/pack)
Specifications
Bottom ShapeFlat
Capacity (Metric)34 mL
DisposableSingle-use
MaterialAluminum
ShapeRound
TypeWeighing Dish
For Use With (Application)For general purpose weighing, dispensing, storage, food processing, evaporating, or drying