Check Order Status
Glass Graduated Class B Type 1 Pipettes
Actual product may vary
Fisherbrand
Glass Graduated Class B Type 1 Pipettes
Pipet liquids securely. Fisherbrand™ graduated class B type 1 pipets are made from soda lime glass. Type 1 pipets have the zero at the top.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Graduations
Tolerance
FKS11902178
5 mL
0.2 mL
±0.20 mL
FKS11912178
10 mL
0.2 mL
±0.20 mL
2 Options
Catalog number
FKS11902178
Price (MYR)
95.00
Add to cart
Volume (Metric):
5 mL
Graduations:
0.2 mL
Tolerance:
±0.20 mL
Price (MYR)
95.00
Add to cart
Soda lime Glass
Blue ceramic graduations
Type 1 Pipet
Class B
Specifications
Certifications/Compliance
Class B, BS 700 ISO 835
Class
B
Material
Soda Lime Glass
Graduated
Yes
Graduations
0.2 mL
Quantity
5 Pack
Tolerance
±0.20 mL
Volume (Metric)
5 mL
Type
Pipet
Unit Size