PTFE Stir Bars with Pivot Ring
PTFE Stir Bars with Pivot Ring
Actual product may vary
Fisherbrand
PTFE Stir Bars with Pivot Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
FKS1189886260 mm10 mm
Catalog number FKS11898862
Price (MYR)
287.00
Add to cart
Length (Metric):
60 mm
Diameter (Metric):
10 mm
Price (MYR)
287.00
Add to cart
Provides stable stirring action. FisherbrandTM PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action.
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Diameter (Metric)10 mm
Length (Metric)60 mm
ShapeCylinder, Pivot ring
Unit Size