Close-it™ Control Seal
Close-it™ Control Seal
Actual product may vary
Close-it™ Control Seal
Prevent sample loss with these adhesive seals. Buerkle™ Close-it™ Control Seals were developed to prevent sample loss through holes created by sampling lances. Labels have a sandwich-type structure (aluminum/paper) and seal containers hermetically (vapor barrier).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHeight (Metric)Width (Metric)dimPrintTechnology
FKS11893352Green95 mm95 mm
FKS11877043Green150 mm150 mm
Catalog number FKS11893352
Price (MYR)
997.00
PK
Add to cart
Color:
Green
Height (Metric):
95 mm
Width (Metric):
95 mm
Price (MYR)
997.00
PK
Add to cart
  • Prevents hygroscopic powders from clumping together
  • Provides quality control information (sampling date, release yes/no, name of tester, etc.)
  • Available in different colors with printing in red, blue, green, yellow, black, neutral, without printing in white
  • Close-it can be written on with ball-point pen, pencil or felt-tip marker
  • The rear of the label is coated with a powerful adhesive
  • Sticks firmly to most commercial containers, even to surfaces coated with fine powder, slightly moist or even frozen surfaces
  • Supplied in handy rolls
  • Cross perforations allow each label to be separated from the roll individually
Also available for food & pharma sector (with certified adhesive)
Specifications
DescriptionClose-it control seal
ColorGreen
MaterialAluminum, Paper
Product TypeControl Seal
Display TextYes
For Use With (Application)Sampling, Used for Providing Quality Control Information (Sampling Date, Release Yes/no, Name of Tester, Etc.)
Quantity500/Roll
Dimensions (LxW)95 x 95 mm
Width (Metric)95 mm
Height (Metric)95 mm
Unit SizePK