Series 3 Model 400W Cooled Incubator, 450 L, Stainless Steel
Series 3 Model 400W Cooled Incubator, 450 L, Stainless Steel
Actual product may vary
Series 3 Model 400W Cooled Incubator, 450 L, Stainless Steel
Temperature Range: -10°C to +50°C
Have Questions?
Catalog NumberCapacity (Metric)
FKS11887542450 L
Catalog number FKS11887542
Price (MYR)
127,238.00
EA
Add to cart
Capacity (Metric):
450 L
Price (MYR)
127,238.00
EA
Add to cart
  • Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
  • Reliable and accurate temperature control
  • Standard single and dual temperature, plus programmable models available
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient control system and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Digital controls are easy to read and simple to use
  • Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications
Certifications/ComplianceISO 9001:2008
Controller TypeStandard digital
DescriptionFreestanding incubator
Capacity (Metric)450 L
Chamber MaterialStainless Steel
Convection TechnologyFan assisted air circulation via premixing chamber
Dimensions (D x W) Shelf510 mm x 600 mm
LightingOptional Fluorescent/UV
MaterialStove Enameled Mild Steel
No. of Shelves3
Temperature Range (Metric)-10°C to +50°C
TypeRefrigerated Incubator
DisplayDigital
InsulationFoamed polyurethane
Model400W
No. of Doors1
RefrigerantCFC-free
Shelf Area (Metric)510 x 600 mm
Temperature Stability±0.1°C
Temperature Uniformity±0.5°C
Warranty2 Years
Width (English)27.1 in.
Width (English) Interior21.6 in.
Width (Metric)690 mm
Width (Metric) Interior550 mm
Height (English)71.2 in.
Height (English) Interior46.1 in.
Height (Metric)1810 mm
Height (Metric) Interior1170 mm
Depth (English) Interior27.6 in.
Depth (Metric) Interior700 mm
Weight (Metric)140 kg
Electrical Requirements240 V, 50 Hz
Voltage230 V
Frequency50 Hz
Unit SizeEA