Check Order Status
Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Actual product may vary
Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Excellent for all media that do not attack rubber. Fisherbrand™ Pipette Bulbs come with three glassball valves, adjustable by finger pressure.
Have Questions?
Change view
Catalog Number
Color
FKS11880950
Green
FKS11890950
Red
2 Options
Catalog number
FKS11880950
Price (MYR)
12.66
EA
Add to cart
Color:
Green
Price (MYR)
12.66
EA
Add to cart
Available in a range of colors
With ca. 60mL volume suitable for pipettes from 2 to 50mL
Pipette media must not enter pipette filler; it may cause valve malfunction
Specifications
Color
Green
Material
Rubber
Type
Pipette Filler
Volume (Metric)
60 mL
For Use With (Equipment)
Can be used on bulb or cylindrical pipettes from 2mL to 50mL
Quantity
1 Each