Ward Pattern Furnace Tongs
Ward Pattern Furnace Tongs
Actual product may vary
Fisherbrand
Ward Pattern Furnace Tongs
For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS11869701740 mm
FKS11899601590 mm
Catalog number FKS11869701
Price (MYR)
490.24
Add to cart
Length (Metric):
740 mm
Price (MYR)
490.24
Add to cart
  • Manufactured from stainless steel
  • For handling a wide variety of crucibles, especially in muffle furnaces
  • Operated by thumb action with one hand
Specifications
For Use With (Application)For handling a wide variety of crucibles and to protect the hand against radiation from the furnace
Quantity1 Ea.
Length (Metric)740 mm
MaterialStainless Steel
TypeFurnace Tongs
Unit Size