Polypropylene Racks for Autosampler Vials
Polypropylene Racks for Autosampler Vials
Actual product may vary
Polypropylene Racks for Autosampler Vials
Polypropylene vial racks are resistant to most solvents.
Have Questions?
Catalog NumberDimensions (L x W x H)For Use With (Equipment)
FKS11767746200 x 105 x 17mm1.5mL vials
Catalog number FKS11767746
Price (MYR)
250.00
Add to cart
Dimensions (L x W x H):
200 x 105 x 17mm
For Use With (Equipment):
1.5mL vials
Price (MYR)
250.00
Add to cart
  • Vial-Rack
  • 50 positions
  • Blue
  • Stackable
  • Feature alphanumeric indexing for easier vial identification
  • Stackable for efficient storage
Specifications
Product TypeVial Rack
Closure IncludedNo
AutoclavableAutoclavable
ColorBlue
MaterialPolypropylene
Positions50
For Use With (Equipment)1.5mL vials
Dimensions (L x W x H)200 x 105 x 17mm
Unit Size