Check Order Status
Grade 113 Cellulose Medium Filtering Qualitative Filter Paper
Actual product may vary
Fisher Catalogue
Grade 113 Cellulose Medium Filtering Qualitative Filter Paper
Filters fine particulates.
Have Questions?
Change view
Catalog Number
Format
Diameter (Metric)
DimensionsLxW
FKS11764193
Disc
150 mm
FKS11780235
Folded Disc
500 mm
2 Options
Catalog number
FKS11764193
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Format:
Disc
Diameter (Metric):
150 mm
Pure grade
Made of cellulose fibers
Alpha content of nearly 100% according precise norms
Ash content:0.06%
Application
Used for liquids, fine particulates
Used in essential, industrial oil
Atmospheric analysis
Beverage industry
Food, chemical industry
Specifications
Description
Ash Content: 0.06%
Format
Disc
Material
Cellulose
Particle Retention
5 to 8 μm
Pore Size
5 to 8 μm
For Use With (Application)
Suitable for fast filtration of liquids with fine particle
Grade
113
Quantity
100 Pk.
Thickness (Metric)
0.16 mm
Diameter (Metric)
150 mm
Unit Size