DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
Actual product may vary
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
Fisherbrand
DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
The gold standard for multifunctional glass laboratory bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11752272500 mL
FKS12347178
FKS1004799020,000 mL
Catalog number FKS11752272
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original GL bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • Retrace Code, and certificate for manufacturing lot traceability
  • Fired-on, highly durable white print
  • Suitable for long-term storage
  • Easy-to-read scale with graduation marks to estimate the content
Specifications
Capacity (Metric)500 mL
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
IncludesWith screw cap and pouring ring from PP (blue)
MaterialBorosilicate Glass (3.3)
Quantity10/Pk.
Thread Size (Metric)GL 45
AutoclavableAutoclavable
Diameter (Metric)86 mm
Unit Size