Clifton™ Nickel 200 Micro Laboratory Spoons
Clifton™ Nickel 200 Micro Laboratory Spoons
Actual product may vary
Fisherbrand
Clifton™ Nickel 200 Micro Laboratory Spoons
Nickel 200 construction maintains product purity in the handling of foods, synthetic fibers, and caustic alkalis. Nickel Electro™ Clifton™ Nickel 200 Micro Laboratory Spoons contain high thermal and electrical conductivities.
Have Questions?
Catalog NumberLength (Metric)
FKS11736617150 mm
Catalog number FKS11736617
Price (MYR)
506.52
Add to cart
Length (Metric):
150 mm
Price (MYR)
506.52
Add to cart
  • Laboratory Spoon
  • Nickel 200 construction - contains minimum 99.6% nickel
  • Bright finish
  • Length: 100mm
  • Spoon measurement: 8mm wide x 6mm long x 1.5mm deep.
  • Spoon capacity (liquid:) 0.2mL
  • Center barrel: diameter 1.60mm
Specifications
Quantity10 Pk.
AutoclavableAutoclavable
Capacity (Metric)0.2 mL
Product TypeLaboratory Spoon
FinishBright
For Use With (Application)For Maintaining Structural Applications
Width (Metric)8 mm
Blade MaterialNickel
Dimensions (L x W)150 x 8 mm
Temperature (Metric) Max315°C
Handle MaterialNickel
MaterialNickel
Length (Metric)150 mm
Unit Size