Stainless Steel Tall Form Container
Stainless Steel Tall Form Container
Actual product may vary
Stainless Steel Tall Form Container
Safely store and transport liquid gases with a container that includes a clamp to secure the lid to the body. Day Impex™ Stainless Steel Tall Form Container house a range of Dewar Flasks and are constructed of stainless steel for anti-corrosive properties.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Outer
FKS117363991 L116mm
FKS117463992 L142mm
Catalog number FKS11736399
Price (MYR)
3,116.00
EA
Add to cart
Capacity (Metric):
1 L
Diameter (Metric) Outer:
116mm
Price (MYR)
3,116.00
EA
Add to cart
  • Include clamp lid, vent, and handle
  • Vacuum insulated
  • Height dimesions include lid
  • Rust-free
  • Anti-corrosive
  • All hygienic
Specifications
Diameter (Metric) Outer116mm
For Use With (Application)For the transport of liquefied gases
Height (Metric)228mm
Height (Metric) Inner185mm
IncludesWith clamp lid attachment, vent and handle
Capacity (Metric)1 L
Depth (Metric) Interior185 mm
Diameter (Metric) Inner85 mm
MaterialStainless Steel
StyleTall Form
Unit SizeEA