Wheaton™ Reusable PVC Coliwasas and Accessories
Wheaton™ Reusable PVC Coliwasas and Accessories
Actual product may vary
Wheaton™ Reusable PVC Coliwasas and Accessories
For a variety of applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Length (Metric)Product Type
FKS117341294 ft.1.21 mExtension and Connector
FKS124373116 ft.1.82 mColiwasa Sampler
Catalog number FKS11734129
Price (MYR)
4,072.00
EA
Add to cart
Length (English):
4 ft.
Length (Metric):
1.21 m
Product Type:
Extension and Connector
Price (MYR)
4,072.00
EA
Add to cart
Specifications
Product TypeExtension and Connector
Capacity (English)25.6 oz.
Capacity (Metric)760 mL
For Use With (Equipment)Clear PVC reusable coliwasas
Length (English)4 ft.
Length (Metric)1.21 m
Unit SizeEA