Grade 53 Ashless Hardened Quantitative Filter Paper
Grade 53 Ashless Hardened Quantitative Filter Paper
Actual product may vary
Fisherbrand
Grade 53 Ashless Hardened Quantitative Filter Paper
Used for quantitative analysis.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11732395125 mm
Catalog number FKS11732395
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
125 mm
  • Plain disc, wet strengthened ashless filter paper
  • Made with 100% cotton linters fibers
  • Paper have been washed out with acid
  • Finally cleaned with demineralized water
  • Ash content: 0.007%
Applications:
  • Used for analysis of acid solution and pressure filtration
  • Food industry
  • Beverage industry
  • Atmospheric analysis
Specifications
DescriptionAsh Content: <0.008%
FormatDisc
MaterialCotton Linters
Particle Retention5 to 8 μm
Pore Size5 to 8 μm
Filtration Speed50 sec.
For Use With (Application)For Acid solutions analysis and for filtration under high pressure
Grade53
Quantity100 Pk.
Thickness (Metric)0.16 mm
Diameter (Metric)125 mm
Unit Size