Nickel Dual Spoon/Spatula Combo Utensils
Nickel Dual Spoon/Spatula Combo Utensils
Actual product may vary
Fisherbrand
Nickel Dual Spoon/Spatula Combo Utensils
Use one tool for two jobs. Nickel Electro™ Nickel Dual Spoon/Spatula Combo Utensils measure liquids and powders with the spoon end and divide and separate materials with the spatula end.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS11726567150 mm
FKS11716567120 mm
FKS11746567210 mm
Catalog number FKS11726567
Price (MYR)
146.00
Add to cart
Length (Metric):
150 mm
Price (MYR)
146.00
Add to cart
Use one tool for two jobs. Nickel Electro™ Nickel Dual Spoon/Spatula Combo Utensils measure liquids and powders with the spoon end and divide and separate materials with the spatula end.
Dual-use utensil measures liquids and fine powders with the spoon end; divides and separates materials with the spatula end.
  • Manufactured from Nickel.
Specifications
Quantity1
Capacity (Metric)2 mL
Product TypeSpatula
Width (Metric)20 mm
Dimensions (L x W)150 x 20 mm
MaterialNickel
Length (Metric)150 mm
Unit Size