Turnover Septum Stoppers
Turnover Septum Stoppers
Actual product may vary
Turnover Septum Stoppers
For narrow-aperture containers
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDiameter (Metric) InnerSizeFor Use With (Equipment)
FKS11716487Red20 mm45 mmFor 2.2 to 2.3cm I.D. containers
FKS11562493Red14 mm29 mmFor 1.5 to 1.6cm I.D. containers
FKS11552493Red12.5 mm25 mmFor 1.3 to 1.35cm I.D. containers
FKS11538132Cream17.5 mm37 mmFor 1.77 to 1.87cm I.D. containers
Catalog number FKS11716487
Price (MYR)
176.00
PK
Add to cart
Color:
Red
Diameter (Metric) Inner:
20 mm
Size:
45 mm
For Use With (Equipment):
For 2.2 to 2.3cm I.D. containers
Price (MYR)
176.00
PK
Add to cart
For narrow-aperture containers
  • External serrations on plug
  • Turnover sleeve folds down over the container's outer neck and creates a secure seal
  • Easily perforated with hypodermic needles
  • Available in red or cream rubber in packs of 10
Specifications
Diameter (Metric) Inner20 mm
For Use With (Application)For narrow aperture containers
For Use With (Equipment)For 2.2 to 2.3cm I.D. containers
Size45 mm
Total Positions1
ColorRed
MaterialRubber
Product TypeTurnover Septum Stopper
Stopper Number4
Unit SizePK