Floating Foam Tube Racks
Floating Foam Tube Racks
Actual product may vary
Fisherbrand
Floating Foam Tube Racks
Float tubes in water baths, tubs or beakers. Fisherbrand™ Floating Foam Tube Racks are constructed of flexible, high-density polyethylene foam for a variety of lab applications. Brightly colored, reusable racks include detachable carrying handles to easily lift samples out of baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsColorShape
FKS117147368 x 15 mL TubesBlueRectangular
FKS1173656518 x 0.2 mL Tubes, 0.5 mL Tubes, and 1.5 to 2.0 mL TubesBlueCircle
FKS117047364 x 50 mL TubesGreenSquare
Catalog number FKS11714736
Price (MYR)
189.00
Add to cart
Holds:
8 x 15 mL Tubes
Color:
Blue
Shape:
Rectangular
Price (MYR)
189.00
Add to cart
  • Brightly colored foam racks are ideal for floating tubes in water baths, tubs or beakers
  • Closed cell foam resists water absorption so racks can be washed and reused
  • Include detachable carrying handles to lift samples easily out of baths
Specifications
AutoclavableNot Autoclavable
ColorBlue
TypeTube Rack
For Use With (Application)For floating tubes in water baths, tubs or beakers
Holds8 x 15 mL Tubes
IncludesDetachable carrying handles
MaterialHigh-density Polyethylene Foam
No. per Pack5 Pk.
ShapeRectangular
Dimensions (L x W x H)17.2 x 5.7 x 6.5 cm
Diameter (Metric) Well13 mm
Unit Size