Check Order Status
Grade 601 Cellulose General Purpose Filter Paper
Actual product may vary
Fisherbrand
Grade 601 Cellulose General Purpose Filter Paper
Used in analytical works in laboratories. Fisherbrand™ Grade 601 Cellulose General Purpose Filter Paper has medium filtration velocity.
Have Questions?
Change view
Catalog Number
Format
Diameter (Metric)
Dimensions (L x W)
FKS11705365
Sheet
570 x 460mm
FKS11765345
Disc
320 mm
2 Options
Catalog number
FKS11705365
Price (MYR)
387.00
Add to cart
Format:
Sheet
Dimensions (L x W):
570 x 460mm
Price (MYR)
387.00
Add to cart
Applications:
Used in hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food industries
Specifications
Format
Sheet
Material
Cellulose
Particle Retention
5 to 13 μm
Pore Size
5 to 13 μm
For Use With (Application)
Filter Paper used in analytical works in laboratories, medium filtration velocity
Grade
601
Quantity
100/Pk.
Thickness (Metric)
0.16 mm
Dimensions (L x W)
570 x 460mm
Length (Metric)
570 mm
Width (Metric)
460 mm
Unit Size