Sterile Cell Strainers
Sterile Cell Strainers
Actual product may vary
Sterile Cell Strainers
Sterile Cell Strainers
Sterile Cell Strainers
Sterile Cell Strainers
Fisherbrand
Sterile Cell Strainers
Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorMesh Size
FKS11587522Blue40 μm
FIS22-363-548White70 μm
FKS11597522White70 μm
CRG352360Yellow100 μm
FIS22-363-549Yellow100 μm
FKS11517532Yellow100 μm
FIS22-363-547Blue40 μm
Catalog number FKS11587522
Price (MYR)
705.00
Add to cart
Color:
Blue
Mesh Size:
40 μm
Price (MYR)
705.00
Add to cart
Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
Rapidly isolate primary cells
  • Fast, simple alternative to gauze filtration
  • Reliably dissociates cells from clumps or primary tissues
  • Consistently obtain a uniform single-cell suspension
  • Ideal for preparation of:
    • Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
    • Specimens for primary cell cultures and immunogens
    • Freezing stocks
    • Filtering agglutinative proteins produced in inactivation serum
Reliable and easy to use
  • Strong nylon mesh with evenly spaced pores for consistent results
  • Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
  • Includes an extended lip for easier handling with forceps
  • Color-coded polypropylene frame with tab enables easy handling and identification
  • Individually packed, ready to use and disposable
Specifications
Mesh MaterialNylon
DisposableSingle-use
ColorBlue
MaterialPolypropylene
Product TypeCell Strainer
SterilitySterile
For Use With (Equipment)Conical Tubes, 50 mL
Mesh Size40 μm
Unit Size