Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size | Height (Metric) | Tolerance |
---|---|---|---|---|
FKS11586923 | 10 mL | 10/19 | 89 mm | ±0.04 mL |
FKS11566933 | 1000 mL | 24/29 | 300 mm | ±0.4 mL |
FKS11526933 | 100 mL | 12/21 | 170 mm | ±0.1 mL |
FKS11576933 | 2000 mL | 29/32 | 370 mm | ±0.6 mL |
FKS11536933 | 200 mL | 14/23 | 210 mm | ±0.15 mL |
FKS11596923 | 20 mL | 10/19 | 111 mm | ±0.04 mL |
FKS11506933 | 25 mL | 10/19 | 110 mm | ±0.04 mL |
FKS11546933 | 250 mL | 14/23 | 220 mm | ±0.15 mL |
FKS11556933 | 500 mL | 19/24 | 260 mm | ±0.25 mL |
FKS11516933 | 50 mL | 12/21 | 139 mm | ±0.06 mL |
FKS11576923 | 5 mL | 10/19 | 69 mm | ±0.025 mL |