Borosillicate Glass Funnel
Borosillicate Glass Funnel
Actual product may vary
Fisherbrand
Borosillicate Glass Funnel
Plain glass funnel manufactured from borosilicate glass. Fisherbrand™ Borosilicate Glass Funnel is ideal for use with funneling liquids in laboratory applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemLength (Metric) Stem
FKS1157242355mm8mm60mm
FKS11592423100mm12mm110mm
FKS1158242375mm9mm80mm
Catalog number FKS11572423
Price (MYR)
348.00
Add to cart
Diameter (Metric)Top:
55mm
Diameter (Metric) Stem:
8mm
Length (Metric) Stem:
60mm
Price (MYR)
348.00
Add to cart
Plain glass funnel manufactured from borosilicate glass.
Specifications
Diameter (Metric)Top55mm
Diameter (Metric) Stem8mm
MaterialBorosilicate Glass
Length (Metric) Stem60mm
Unit Size