Eutech™ EC620132 pH electrode
Eutech™ EC620132 pH electrode
Actual product may vary
Eutech™ EC620132 pH electrode
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Have Questions?
Catalog NumberConnector Types
FKS11569074BNC
Catalog number FKS11569074
Price (MYR)
2,007.00
EA
Add to cart
Connector Types:
BNC
Price (MYR)
2,007.00
EA
Add to cart
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Features:
  • Range: 0 to 14 pH
  • Temperature Range: 0 to 60 °C
  • Liquid Junction Type: Open pore
  • Internal Reference Type: Polymer-gel
  • Sealed/Refillable: Sealed
  • Reference Junction: Double
  • Dimensions (Shaft): 105 x 12 mm
  • Cable Length: 1 m
  • Connector: BNC
Specifications
DescriptionOpen pore glass-body pH electrode for general pH measurements
Connector TypesBNC
JunctionDouble
MaterialGlass Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 60°C
TypepH Electrode
For Use With (Application)pH measurement
For Use With (Equipment)All pH meters with BNC input connector
Temperature (Metric) Range0°C to 60°C
Unit SizeEA