Stainless Steel Test Sieve with Woven Wire Mesh
Stainless Steel Test Sieve with Woven Wire Mesh
Actual product may vary
Fisherbrand
Stainless Steel Test Sieve with Woven Wire Mesh
Made from Stainless Steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has a 200mm diameter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore Size
FKS1155756220 μm
FKS115375421.00 mm
FKS11567552106 μm
FKS1150756253 μm
FKS115175421.40 mm
Catalog number FKS11557562
Price (MYR)
4,004.00
Add to cart
Pore Size:
20 μm
Price (MYR)
4,004.00
Add to cart
  • 200mm diameter test sieves
  • Stainless Steel frame
  • Woven wire mesh
  • Supplied with Certificate of Conformity and record card
Specifications
Quantity1 Each
Pore Size20 μm
TypeSieve
Certifications/ComplianceCertificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
MaterialStainless Steel
IncludesCertificate of conformity and record card
Diameter (Metric) Outer200 mm
Frame MaterialStainless Steel
Mesh MaterialWoven Wire
Unit Size