Octahedral PTFE Stir Bar
Octahedral PTFE Stir Bar
Actual product may vary
Fisherbrand
Octahedral PTFE Stir Bar
Provides stable stirring action. These PTFE magnetic stir bars are autoclavable and include a pivot ring. The flat surfaces provide a great mixing action.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
FKS1154778251 mm9.5 mm
Catalog number FKS11547782
Price (MYR)
282.00
Add to cart
Length (Metric):
51 mm
Diameter (Metric):
9.5 mm
Price (MYR)
282.00
Add to cart
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action
  • All have pivot rings
  • AlNiCo-V magnets encased in PTFE
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Diameter (Metric)9.5 mm
Length (Metric)51 mm
Quantity10/Pk.
ShapeOctahedral
Unit Size