Stainless Steel Chattaway Light PTFE Coated Spatulas
Stainless Steel Chattaway Light PTFE Coated Spatulas
Actual product may vary
Stainless Steel Chattaway Light PTFE Coated Spatulas
Get superior results when you use the stainless steel PTFE-coated spatula for your essential laboratory work. Ideal for Biology, Microbiology and Pharmaceutical Applications, the Fisherbrand™ Stainless Steel Chattaway Spatula Light PTFE Coated is widely used, sturdy, affordable, and resistant to most solvents.
Have Questions?
Catalog NumberLength (Metric)
FKS11543502150 mm
Catalog number FKS11543502
Price (MYR)
28.62
EA
Add to cart
Length (Metric):
150 mm
Price (MYR)
28.62
EA
Add to cart
  • Resistant to deterioration from contact with boiling water, acids, bases, and most solvents
  • Ideal for use with a wide range of compounds
  • Ideal for Biology, Microbiology and Pharmaceutical Applications
  • Widely used, sturdy, and affordable
  • One end flat, other end at 45° angle
  • Length = 150mm
  • Blade width = 8mm
  • Centre barrel diameter 2.95mm - 'light'
Specifications
Quantity1 Ea.
CoatingPTFE Xylan™
Product TypeSpatula
For Use With (Application)Ideal for Biology, Microbiology and Pharmaceutical Applications
Width (Metric) Blade8 mm
Temperature (Metric) Max-195°C to +260°C
MaterialStainless Steel
Length (Metric)150 mm
Unit SizeEA