Check Order Status
Solid Rubber Stoppers
Actual product may vary
Fisherbrand
Solid Rubber Stoppers
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
Have Questions?
Catalog Number
Diameter (Metric) Bottom
Stopper Number
Height (Metric)
Diameter (Metric) Top
FKS11542922
15 mm
15
24mm
18 mm
Catalog number
FKS11542922
Price (MYR)
91.00
Add to cart
Diameter (Metric) Bottom:
15 mm
Stopper Number:
15
Height (Metric):
24mm
Diameter (Metric) Top:
18 mm
Price (MYR)
91.00
Add to cart
Offers higher resilience, flexibility and can withstand many chemical agents in normal laboratory use.
Temperature range -40° to +70°C
Not suitable for use with petroleum oils and hydrocarbon solvents
Specifications
For Use With (Application)
Laboratory use
Height (Metric)
24mm
Quantity
20/Pk.
Color
Red
Diameter (Metric) Bottom
15 mm
Diameter (Metric) Top
18 mm
Material
Rubber
Product Type
Solid Stopper
Stopper Number
15
Unit Size