Stainless Steel Trulla Spatulas
Stainless Steel Trulla Spatulas
Actual product may vary
Fisherbrand
Stainless Steel Trulla Spatulas
Designed using high quality stainless steel components these trulla spatulas are ideal in most laboratory, research, and other environments.
Have Questions?
Catalog NumberDimensions (L x W)
FKS11523492176 x 20 mm
Catalog number FKS11523492
Price (MYR)
159.00
Add to cart
Dimensions (L x W):
176 x 20 mm
Price (MYR)
159.00
Add to cart
  • Spatulas, nickel and stainless steel Chattaway 3.6mm barrel, nickel and stainless micros 1.6mm barrel
  • Uses the finest in corrosion-resistant 304 stainless steel
  • Supplied in standard packs of 5
  • For laboratory metalware consumables
  • Designed for corrosion resistance and durability ideal in laboratory routines
Specifications
Quantity5 Pk.
Capacity (Metric)176 mL
Product TypeTrulla Spatula
Width (Metric)20 mm
Diameter (Metric) Outer3.6 mm
Dimensions (L x W)176 x 20 mm
MaterialStainless Steel
Length (Metric)176 mm
Unit Size