Borosilicate Glass Class A Graduated Cylinder
Borosilicate Glass Class A Graduated Cylinder
Actual product may vary
Borosilicate Glass Class A Graduated Cylinder
Borosilicate Glass Class A Graduated Cylinder
Borosilicate Glass Class A Graduated Cylinder
Borosilicate Glass Class A Graduated Cylinder
Measure liquids with precision in glass class A graduated cylinders. Fisherbrand™ borosilicate class A graduated cylinders have a blue ceramic scale and a hexagonal base.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS1151783210 mL0.2 mL
FKS115778321000 mL10 mL
FKS11547832100 mL1 mL
FKS11557832250 mL2 mL
FKS1152783225 mL0.5 mL
FKS11567832500 mL5 mL
FKS1153783250 mL1 mL
Catalog number FKS11517832
Price (MYR)
157.00
Add to cart
Capacity (Metric):
10 mL
Graduations:
0.2 mL
Price (MYR)
157.00
Add to cart
Measure liquids with precision in glass class A graduated cylinders. Fisherbrand™ borosilicate class A graduated cylinders have a blue ceramic scale and a hexagonal base.
  • DIN 12680 BS 604 ISO 4788
  • Graduated in blue ceramic
  • Hexagonal base
  • Individual reorder code on each item
  • Batch Certificate of Compliance' is downloadable 
Specifications
Base TypeHexagonal
Scale ColorBlue
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
ClassClass A
Height (Metric)135 mm
TypeGraduated Cylinder
MaterialBorosilicate Glass
Diameter (Metric) Outside15 mm
Capacity (Metric)10 mL
IncludesConformity Certificate
ScaleBlue Ceramic
Graduations0.2 mL
AutoclavableAutoclavable
Limit of Error±0.1 mL
Unit Size