Made from Stainless Steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has a 200mm diameter.
Have Questions?
Change view
Catalog Number
Pore Size
FKS11517542
1.40 mm
FKS11537542
1.00 mm
FKS11567552
106 μm
FKS11507562
53 μm
FKS11557562
20 μm
5 Options
Catalog number FKS11517542
Price (MYR)
618.00
Add to cart
Pore Size:
1.40 mm
Price (MYR)
618.00
Add to cart
200mm diameter test sieves
Stainless Steel frame
Woven wire mesh
Supplied with Certificate of Conformity and record card
Specifications
Quantity1 Each
Pore Size1.40 mm
TypeSieve
Certifications/ComplianceCertificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000