Made from Stainless Steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has a 200mm diameter.
Have Questions?
Change view
Catalog Number
Pore Size
FKS11507562
53 μm
FKS11537542
1.00 mm
FKS11567552
106 μm
FKS11557562
20 μm
FKS11517542
1.40 mm
5 Options
Catalog number FKS11507562
Price (MYR)
645.00
Add to cart
Pore Size:
53 μm
Price (MYR)
645.00
Add to cart
200mm diameter test sieves
Stainless Steel frame
Woven wire mesh
Supplied with Certificate of Conformity and record card
Specifications
Quantity1 Each
Pore Size53 μm
TypeSieve
Certifications/CompliancePart 1 : 2000, ISO 3310 : Part 1 : 2000, BS410