Get optimum results when you use the nickel-based micro spatulas for your essential laboratory work. Ideal for Biology, Microbiology and Pharmaceutical Applications, the Fisherbrand™ Nickel Chattaway Micro Spatulas are widely used, sturdy, affordable, and resistant to most solvents.
Have Questions?
Catalog Number
Length (Metric)
FKS11503482
150 mm
Catalog number FKS11503482
Price (MYR)
360.00
Add to cart
Length (Metric):
150 mm
Price (MYR)
360.00
Add to cart
Excellent resistance to many corrosive environments
High thermal and electrical conductivities
Low gas content and low vapor pressure
Nickel 200 spec (minimum 99.6% nickel content)
Bright finish
Spatula Blade One end flat, other end at 45° angle