Sieve Brush
Sieve Brush
Actual product may vary
Fisherbrand
Sieve Brush
A double-ended brush for use with Fisherbrand™ Stainless Steel Test Sieves.
Have Questions?
Catalog NumberSize
FKS1150276320 x 230 mm
Catalog number FKS11502763
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
20 x 230 mm
  • Provides gentle brushing
  • Manufactured from either brass or nylon and bristle
  • Two different sizes: 30 x 135mm and 20 x 230mm
Specifications
Product TypeTerm Brush
Bristle MaterialNylon
Handle MaterialBrass
Quantity1 Ea.
Size20 x 230 mm
Unit Size