Check Order Status
Grade 601 General Purpose Filter Papers
Actual product may vary
Fisherbrand
Grade 601 General Purpose Filter Papers
A filter paper used in analytical works in laboratories with medium filtration velocity.
Have Questions?
Change view
Catalog Number
Diameter (Metric)
FKS11445248
185 mm
FKS11572003
42.5 mm
FKS11566873
90 mm
FKS11576873
110 mm
FKS11425248
125 mm
FKS11435248
150 mm
FKS11592003
240 mm
7 Options
Catalog number
FKS11445248
Price (MYR)
94.00
Add to cart
Diameter (Metric):
185 mm
Price (MYR)
94.00
Add to cart
73g/m
2
weight
0.16mm thick
Pore size of 5 to 13μm
Filtration velocity of 88 sec
Burst strength of 1.95kg/cm
2
Available in a range of sizes
Specifications
Format
Disc
Pore Size
5 to 13 μm
Filtration Speed
88 seconds
For Use With (Application)
General works, hospitals, research laboratories, schools, chemical industries, cosmetics, food and beverage industries
Quantity
100/Pk.
Thickness (Metric)
0.16 mm
Diameter (Metric)
185 mm
Unit Size