Interchangeable Block for QB Series Dry Block Heaters
Interchangeable Block for QB Series Dry Block Heaters
Actual product may vary
Fisherbrand
Interchangeable Block for QB Series Dry Block Heaters
For use with 0.2mL tubes, strips and 96-well microtitre plates used in molecular biology and biotechnology applications. Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters can withstand temperatures up to 100°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube SizeNo. of PlacesDepth (Metric)
FKS1143091816mm1250 mm hole depth
GRAQB-17H17mm1075 mm hole depth
GRAQB-1313mm1250 mm hole depth
GRAQB-1010mm2450 mm hole depth
GRAQB-1212mm2450 mm hole depth
Catalog number FKS11430918
Price (MYR)
1,836.00
Add to cart
Tube Size:
16mm
No. of Places:
12
Depth (Metric):
50 mm hole depth
Price (MYR)
1,836.00
Add to cart
  • For use with QBD1, QBD2, QBD4, QBH2, QBA1 and QBA2 dry block heating system
  • Accommodates a variety of test tube sizes
  • Available in plain block format without holes
Specifications
For Use With (Equipment)Test Tubes (16 mm)
MaterialAluminum
No. of Places12
TypeBlock
Tube Size16mm
Dimensions (L x W x H)140 x 50 x 63 mm
Depth (Metric)50 mm hole depth
Unit Size