MBL™ Graduated Class AS Type 1 Glass Pipets
MBL™ Graduated Class AS Type 1 Glass Pipets
Actual product may vary
Fisherbrand
MBL™ Graduated Class AS Type 1 Glass Pipets
Pipet with precision. MBL™ class AS type 1 soda-lime glass pipets have graduations in permanent amber stain. Type 1 pipets have the zero mark at the top of the pipet.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)GraduationsColorTolerance
FKS1138965310 mL0.1 mLOrange±0.050 mL
FKS1139965325 mL0.1 mLGreen±0.10 mL
FKS113596531 mL0.01 mLYellow±0.007 mL
FKS113696532 mL0.02 mLBlack±0.01 mL
FKS113796535 mL0.05 mLRed±0.03 mL
Catalog number FKS11389653
Price (MYR)
134.00
Add to cart
Volume (Metric):
10 mL
Graduations:
0.1 mL
Color:
Orange
Tolerance:
±0.050 mL
Price (MYR)
134.00
Add to cart
  • Complies with ISO 835
  • Calibrated to deliver from zero at the top to any graduation line down to the shoulder
  • Graduations in permanent amber stain which resists aggressive washing solutions
  • Each pipette is calibrated to deliver with a +5s waiting time
  • Manufactured from soda lime glass with tooled jets for extra strength and to ensure correct outflow times
Specifications
CalibratedYes
Certifications/ComplianceISO 835
MaterialSoda Lime Glass
GraduatedYes
Graduations0.1 mL
Tolerance±0.050 mL
ColorOrange
Volume (Metric)10 mL
TypePipet
Unit Size