Whatman™ 602H 1/2 Qualitative Folded Filter Paper Circle
Whatman™ 602H 1/2 Qualitative Folded Filter Paper Circle
Actual product may vary
Whatman™ 602H 1/2 Qualitative Folded Filter Paper Circle
Dense filter paper forr collecting and removing fine precipitates
Have Questions?
Catalog NumberDiameter (Metric)
FKS1137524490 mm
Catalog number FKS11375244
Price (MYR)
540.00
PK
Add to cart
Diameter (Metric):
90 mm
Price (MYR)
540.00
PK
Add to cart
  • Used in sample preparation
  • Residual sugar determination
  • Acidic spectra
  • Refractometric analysis
  • Slow flow speed
  • HPLC
Specifications
DescriptionGrade 602 h ½ Qualitative Filter Paper Folded (Prepleated), 90 mm
FormatPre-pleated Circle
MaterialCellulose
Particle Retention<2 μm
Filtration SpeedApprox. 375 s
For Use With (Application)A dense prepleated filter paper for collecting very small particles and removing fine precipitates.
GradeGrade 602 h ½
Diameter (Metric)90 mm
Unit SizePK