Sterilin™ Plastic Transfer Pipettes
Sterilin™ Plastic Transfer Pipettes
Actual product may vary
Fisherbrand
Sterilin™ Plastic Transfer Pipettes
Transfer liquids safely and accurately with Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes. Ideal for general laboratory use, sample storage and transport.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Quantity
FKS113692033 mL10/Pk., 500/Cs.
FKS129094873 mL500/Pk., 3000/Cs.
Catalog number FKS11369203
Price (MYR)
304.84
Add to cart
Volume (Metric):
3 mL
Quantity:
10/Pk., 500/Cs.
Price (MYR)
304.84
Add to cart
Transfer liquids safely and accurately with Thermo Scientific™ Sterilin™ Plastic Transfer Pipettes. Ideal for general laboratory use, sample storage and transport.
  • Manufactured from nontoxic low density polyethylene
  • Integral filling bulb eliminates contamination associated with separate rubber bulbs
  • Excellent transparency and uniform thickness
  • Precise graduations on 1 and 3mL versions ensuring consistent results
  • Can be heat sealed for sample storage and transportation
  • Heat sealed bulbs suitable for storage in the gas phase of liquid nitrogen
  • 3mL graduated: suitable for blood banking and general laboratory use
  • Narrow stem: suitable for use as a freeze vial, sedimentation and transport
  • 1mL graduated: suitable for general laboratory use, storage and transportation
  • of samples
Ordering Information
  • Not available in all geographic regions
Specifications
MaterialLDPE
SterilitySterile
GraduatedYes
Quantity10/Pk., 500/Cs.
Volume (Metric)3 mL
Unit Size