Azlon™ Polypropylene Graduated Molded Jugs with Handles
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Graduated Molded Jugs with Handles
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Graduations
FKS11353694
2000 mL
500mL
FKS11333694
500 mL
100mL
FKS11343694
1000 mL
100mL
FKS11373694
5000 mL
500mL
4 Options
Catalog number FKS11353694
Price (MYR)
55.00
Add to cart
Capacity (Metric):
2000 mL
Graduations:
500mL
Price (MYR)
55.00
Add to cart
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
Tapered jugs in high clarity polypropylene
Permanent molded graduations
Autoclavable at 121° C
Ergonomically designed handle with thumb grip for easy carrying