Azlon™ Polypropylene Graduated Molded Jugs with Handles
Azlon™ Polypropylene Graduated Molded Jugs with Handles
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Graduated Molded Jugs with Handles
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS113436941000 mL100mL
FKS11333694500 mL100mL
FKS113536942000 mL500mL
FKS113736945000 mL500mL
Catalog number FKS11343694
Price (MYR)
31.00
Add to cart
Capacity (Metric):
1000 mL
Graduations:
100mL
Price (MYR)
31.00
Add to cart
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
  • Tapered jugs in high clarity polypropylene
  • Permanent molded graduations
  • Autoclavable at 121° C
  • Ergonomically designed handle with thumb grip for easy carrying
Specifications
Capacity (Metric)1000 mL
Capacity (English)33.81 oz.
ColorClear
MaterialPolypropylene
No. per PackIndividual
QuantityEach
Temperature Range (Metric)–20 to 135°C
Graduations100mL
Certifications/ComplianceEU; USA
Closure IncludedNo
AutoclavableAutoclavable
GraduatedYes
MouthWide
ShapeTapered
Height (Metric)180 mm
Product TypeMolded Jug
Height (English)7.09 in.
Diameter (Metric)104 mm
Through the Closure DispensingNo
Unit Size