Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size | Tolerance |
---|---|---|---|
FKS11339443 | 250 mL | 14/23 | ±0.15 mL |
FKS11359443 | 1000 mL | 24/29 | ±0.4 mL |
FKS11399433 | 25 mL | 10/19 | ±0.04 mL |
FKS11349443 | 500 mL | 19/24 | ±0.25 mL |
FKS11309443 | 50 mL | 12/21 | ±0.06 mL |
FKS11319443 | 100 mL | 12/21 | ±0.01 mL |