Azlon™ Multi-lingual Safety Venting Wash Bottle
Azlon™ Multi-lingual Safety Venting Wash Bottle
Actual product may vary
Fisherbrand
Azlon™ Multi-lingual Safety Venting Wash Bottle
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Chemical LabelDescriptionClosure Color
FKS11303524500 mLIsopropanolPrinted in English, French, German and SpanishBlue
FKS12196220250 mLIsopropanolPrinted in English, French, German and SpanishBlue
FKS12357512500 mLEthanolRegion 2: Norwegian, Danish, Swedish, EnglishOrange
FKS12550456500 mLMethanolPrinted in English, French, German and SpanishGreen
Catalog number FKS11303524
Price (MYR)
511.00
Add to cart
Capacity (Metric):
500 mL
Chemical Label:
Isopropanol
Description:
Printed in English, French, German and Spanish
Closure Color:
Blue
Price (MYR)
511.00
Add to cart
Store and use a variety of solvents with multi-lingual labels for improved safety. Azlon Multi-lingual LDPE Venting Wash Bottles are available in four different multi-language labeling combinations. Solvent content labels correspond to color-coded lids for easy identification in a variety of settings.
  • Multi-lingual wide neck wash bottles featuring the patented DripLok™ vapor-venting valve that prevents solvent drips
  • Indelibly printed in multiple languages (based on region)
  • Includes the Globally Harmonised Symbols, NFPA diamond, risk phrases, and safety information
  • Fitted with color coded one piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)500 mL
Capacity (English)16.91 oz.
ColorTranslucent
DescriptionPrinted in English, French, German and Spanish
IncludesPolypropylene caps and colored beaks with fine nozzle end to control the jet accurately
MaterialLDPE
Length (Metric)33.07cm
Quantity5/PK
Temperature Range (Metric)–50 to 80°C
Width (Metric)19.08cm
Closure ColorBlue
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthWide
ShapeCylindrical
Height (Metric)5.6 cm
Product TypeMulti-Lingual Printed Wash Bottle
Chemical LabelIsopropanol
Closure MaterialPolypropylene
Closure TypeCap
Through the Closure DispensingYes
Unit Size