Eutech™ pH Buffers and Solutions
Product Image
Eutech™ pH Buffers and Solutions
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes
Have Questions?
Catalog NumberpH
FKS107595031.68
Catalog number FKS10759503
Price (MYR)
161.00
EA
Add to cart
pH:
1.68
Price (MYR)
161.00
EA
Add to cart
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
DescriptionBuffer Solution
For Use With (Equipment)pH Meter and Electrode
pH1.68
Quantity480 mL
Solution TypeBuffer
Unit SizeEA