SILBERBRAND™ Amber Borosilicate Glass Automatic Burette
SILBERBRAND™ Amber Borosilicate Glass Automatic Burette
Actual product may vary
SILBERBRAND™ Amber Borosilicate Glass Automatic Burette
Includes intermediate stopcock with PTFE key. BRAND™ SILBERBRAND™ Amber Glass Automatic Burette is calibrated to deliver.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1065869150 mL
Catalog number FKS10658691
Price (MYR)
4,898.00
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
4,898.00
Add to cart
  • Borosilicate glass 5.4
  • Pellet pattern
  • Error limits according to Class B, DIN EN ISO 385
  • Interchangeable keys
  • Calibrated to deliver (TD, Ex)
  • With glass-key in the titration and intermediate stopcock to recirculate residual liquid
  • Automatic zeroing
Specifications
ColorAmber
Capacity (Metric)50 mL
For Use With (Equipment)Titration Stopcock With PTFE Key or STJ Glass Stopcock
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesGlass Bottle
ScaleAmber
Stopcock MaterialPTFE Key
Tolerance±0.1 mL
TypeAutomatic Buret
ClassClass B
Certifications/ComplianceDIN EN ISO 385
Unit Size